Stampa…

D-Homophenylalanine 98%

Fornitore: Thermo Scientific

Synonyms: (R)-2-Amino-4-phenylbutyric Acid, H-D-HomoPhe-OH, H-D-HoPhe-OH

H27629.06 H27629.03
H27629.06EA 471 EUR
H27629.06 H27629.03
D-Homophenylalanine 98%
D-Homophenylalanine

Purity: 98% CAS No.: 82795-51-5 Molecular Formula: C10H13NO2 Formula Weight: 179.22 Melting Point: 300° Beilstein Reference: 4675530 MDL No.: MFCD00063091

Formula: C₁₀H₁₃NO₂
Peso molecolare: 179,22 g/mol
Punto di fusione: >300 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00063091
CAS Number: 82795-51-5

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR