(S)-2-Amino-6-guanidinohexanoic acid hydrochloride ≥98%
Fornitore: Thermo Scientific
Synonyms:
H-HoArg-OH·HCl, L(+)-Homoarginine hydrochloride, H-HomoArg-OH.HCl, H-HomoArg-OH·HCl
Purity: 98+%
CAS No.: 1483-01-8
Molecular Formula: C7H16N4O2.HCl
Formula Weight: 224.69
Melting Point: 213-215°
Beilstein Reference: 4016621
Optical Rotation: +18° (c=1 in 1N HCl)
MDL No.: MFCD00012622
Formula:
C₇H₁₆N₄O₂·HCl Peso molecolare: 224,7 g/mol Punto di fusione: 213…215 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00012622 CAS Number: 1483-01-8 EINECS: 216-045-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...