Stampa…

p-Sexiphenyl 95%

Fornitore: Thermo Fisher Scientific

H26586.MD H26586.03
H26586.MDEA 169 EUR
H26586.MD H26586.03
p-Sexiphenyl 95%
p-Sexiphenyl

Purity: 95% CAS No.: 4499-83-6 Molecular Formula: C36H26 Formula Weight: 458.59 Melting Point: 400° MDL No.: MFCD00039559

Formula: C₃₆H₂₆
Punto di fusione: >400 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00039559
CAS Number: 4499-83-6
EINECS: 224-799-2

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR