Stampa…

Methyl-2-amino-2-methylpropanoate hydrochloride 99%

Fornitore: Thermo Fisher Scientific

H26550.06 H26550.03
H26550.06EA 236 EUR
H26550.06 H26550.03
Methyl-2-amino-2-methylpropanoate hydrochloride 99%
Methyl-2-amino-2-methylpropanoate hydrochloride

Purity: 99% CAS No.: 15028-41-8 Molecular Formula: C5H11NO2.HCl Formula Weight: 153.61 Storage Temperature: 2°C to 8°C Melting Point: ca 185° dec. MDL No.: MFCD00214247

Formula: C₅H₁₁NO₂·HCl
Punto di fusione: ∼ 185 °C (dec.)
MDL Number: MFCD00214247
CAS Number: 15028-41-8

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR