3,3-Difluoroazetidine hydrochloride 95%
Fornitore: Thermo Scientific
|
Attenzione
|
Synonyms:
3,3-Difluoroazetidine HCl
H25751.03
H25751.MD
H25751.03EA
500
EUR
H25751.03
H25751.MD
3,3-Difluoroazetidine hydrochloride 95%
3,3-Difluoroazetidine hydrochloride
Purity: 95%
CAS No.: 288315-03-7
Molecular Formula: C3H5F2N.HCl
Formula Weight: 129.54
Melting Point: 138-140°
MDL No.: MFCD05663714
Formula:
C₃H₆ClF₂N Peso molecolare: 129,54 g/mol Punto di fusione: 138…140 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD05663714 CAS Number: 288315-03-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...