Stampa…
Cyclobutylamine hydrochloride 97%
Fornitore: Thermo Scientific
Attenzione
Synonyms: Cyclobutanamine hydrochloride
B25400.14 B25400.06
B25400.14EA 277 EUR
B25400.14 B25400.06
Cyclobutylamine hydrochloride 97%
Cyclobutylamine hydrochloride
Purity: 97% CAS No.: 6291-01-6 Molecular Formula: C4H9N.HCl Formula Weight: 107.58 Melting Point: 180-183° MDL No.: MFCD00034953
Formula: C₄H₉N·HCl
Peso molecolare: 107,58 g/mol
Punto di fusione: 180…183 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00034953
CAS Number: 6291-01-6
EINECS: 228-540-4