4-Azabenzimidazole 99%
Fornitore: Thermo Scientific
Synonyms:
4H-Imidazo[4,5-b]pyridine
B25051.03
B25051.06
B25051.14
B25051.03EA
57.2
EUR
B25051.03
B25051.06
B25051.14
4-Azabenzimidazole 99%
4-Azabenzimidazole
Purity: 99%
CAS No.: 273-21-2
Molecular Formula: C6H5N3
Formula Weight: 119.13
Melting Point: 145-150°
Boiling Point: 140-150°/0.1mm
Beilstein Reference: 2600
MDL No.: MFCD00005579
Formula:
C₆H₅N₃ Peso molecolare: 119,127 g/mol Punto di ebollizione: 140…150 °C (0,1 mmHg) Punto di fusione: 145…150 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00005579 CAS Number: 273-21-2 EINECS: 205-987-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...