Stampa…

2-Propionylpyrrole 99%

Fornitore: Thermo Fisher Scientific
Attenzione

B23954.03 B23954.06
B23954.03EA 39.5 EUR
B23954.03 B23954.06
2-Propionylpyrrole 99%
2-Propionylpyrrole

Purity: 99% CAS No.: 1073-26-3 Molecular Formula: C7H9NO Formula Weight: 123.15 Melting Point: 48-51° MDL No.: MFCD01696449

Formula: C₇H₉NO
Punto di fusione: 48…51 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD01696449
CAS Number: 1073-26-3
EINECS: 214-026-7

Visualizza i codici

Specifiche

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR