Stampa…

2-Chloro-4H-1,3,2-benzodioxaphosphorin-4-one 97%

Fornitore: Thermo Scientific
Pericolo

Synonyms: Van Boom's Reagent, Salicylchlorophosphite

B21086.06 B21086.14
B21086.06EA 48.2 EUR
B21086.06 B21086.14
2-Chloro-4H-1,3,2-benzodioxaphosphorin-4-one 97%
2-Chloro-4H-1,3,2-benzodioxaphosphorin-4-one

Purity: 97% CAS No.: 5381-99-7 Molecular Formula: C7H4ClO3P Formula Weight: 202.54 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Melting Point: 36-40° Boiling Point: 127-128°/11mm UN No.: UN3261 MDL No.: MFCD00013353

Formula: C₇H₄ClO₃P
Peso molecolare: 202,53 g/mol
Punto di ebollizione: 127…128 °C (11 mmHg)
Punto di fusione: 36…40 °C
Flash Pt: >110 °C (230 °F)
MDL Number: MFCD00013353
CAS Number: 5381-99-7
ADR: 8,II

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR