Stampa…

Hyodeoxycholic acid 96%

Fornitore: Thermo Scientific
Attenzione

B20506.06 B20506.14 B20506.22
B20506.06EA 79.3 EUR
B20506.06 B20506.14 B20506.22
Hyodeoxycholic acid 96%
Hyodeoxycholic acid

Purity: 96% CAS No.: 83-49-8 Molecular Formula: C24H40O4 Formula Weight: 392.59 Melting Point: 197-199° Optical Rotation: +8° (c=2 in ethanol) MDL No.: MFCD00003681

Formula: C₂₄H₄₀O₄
Peso molecolare: 392,58 g/mol
Punto di fusione: 197…199 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00003681
CAS Number: 83-49-8
EINECS: 201-483-2
Merck Index: 14,4857

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR