Ethyl diacetoacetate 97%
Fornitore: Thermo Scientific
|
Attenzione
|
Synonyms:
Diacetoacetic Acid Ethyl Ester, Ethyl 2-Acetyl-3-oxobutyrate, Ethyl 2-acetyl-3-oxobutanoate
Purity: 97%
CAS No.: 603-69-0
Molecular Formula: C8H12O4
Formula Weight: 172.18
Flash Point: 85°(185°F)
Refractive Index: 1.4720
Boiling Point: 104-105°/17mm
Beilstein Reference: 775014
MDL No.: MFCD00009864
Formula:
C₈H₁₂O₄ Peso molecolare: 172,18 g/mol Punto di ebollizione: 104…105 °C (17 mmHg) Densità: 1,104 Flash Pt: 85 °C (185 °F) Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00009864 CAS Number: 603-69-0 EINECS: 210-053-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...