Stampa…

Methyl-4-chlorocinnamate 99%

Fornitore: Thermo Scientific
Attenzione

Synonyms: Methyl 3-(4-chlorophenyl)acrylate, 4-Chlorocinnamic Acid Methyl Ester

A17071.06 A17071.14
A17071.06EA 60.9 EUR
A17071.06 A17071.14
Methyl-4-chlorocinnamate 99%
Methyl-4-chlorocinnamate

Purity: 99% CAS No.: 7560-44-3 Molecular Formula: C10H9ClO2 Formula Weight: 196.64 Melting Point: 74-77° Beilstein Reference: 2045629 MDL No.: MFCD00075790

Formula: C₁₀H₉ClO₂
Peso molecolare: 196,63 g/mol
Punto di fusione: 74…77 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00075790
CAS Number: 7560-44-3
EINECS: 231-451-3

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR