Stampa…

α-Isopropylphenylacetic acid 97%

Fornitore: Thermo Fisher Scientific
Attenzione

A16077.06 A16077.14
A16077.06EA 93.2 EUR
A16077.06 A16077.14
α-Isopropylphenylacetic acid 97%
alpha-Isopropylphenylacetic acid

Purity: 97% CAS No.: 3508-94-9 Molecular Formula: C11H14O2 Formula Weight: 178.23 Melting Point: 57-62° Beilstein Reference: 2045915 MDL No.: MFCD00021708

Formula: C₁₁H₁₄O₂
Punto di fusione: 57…62 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00021708
CAS Number: 3508-94-9

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR