2-(Phosphonooxy)benzoic acid 98%
Fornitore: Thermo Scientific
|
Attenzione
|
Synonyms:
Fosfosal, 2-Carboxyphenyl Phosphate
A14417.06
A14417.03
A14417.06EA
260
EUR
A14417.06
A14417.03
2-(Phosphonooxy)benzoic acid 98%
2-(Phosphonooxy)benzoic acid
Purity: 98%
CAS No.: 6064-83-1
Molecular Formula: C7H7O6P
Formula Weight: 218.10
Melting Point: 162-166°
Beilstein Reference: 2619142
MDL No.: MFCD00042645
Formula:
C₇H₇O₆P Peso molecolare: 218,10 g/mol Punto di fusione: 162…166 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00042645 CAS Number: 6064-83-1 EINECS: 227-993-5 Merck Index: 14,4255 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...