2,6-Lutidine-N-oxide 98%
Fornitore: Thermo Scientific
|
Attenzione
|
Synonyms:
2,6-Dimethylpyridine N-Oxide
A14299.09
A14299.18
A14299.30
A14299.09EA
48
EUR
A14299.09
A14299.18
A14299.30
2,6-Lutidine-N-oxide 98%
2,6-Lutidine-N-oxide
Purity: 98%
CAS No.: 1073-23-0
Molecular Formula: C7H9NO
Formula Weight: 123.16
Refractive Index: 1.5685
Boiling Point: 108-111°/10mm
Beilstein Reference: 110551
MDL No.: MFCD00039715
Formula:
C₇H₉NO Peso molecolare: 123,16 g/mol Punto di ebollizione: 108…111 °C (10 mmHg) Densità: 1,102 Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00039715 CAS Number: 1073-23-0 EINECS: 214-025-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...