3-Amino-4-pyrazolecarbonitrile 98%
Fornitore: Thermo Fisher Scientific
|
Attenzione
|
Synonyms:
5-Aminopyrazole-4-carbonitrile, 3-Amino-1H-pyrazole-4-carbonitrile, 3-Aminopyrazole-4-carbonitrile, 3-Amino-4-cyanopyrazole 3-Amino-4-pyrazolecarbonitrile 5-Amino-4-cyanopyrazole 3-Amino-1H-pyrazole-4-carbonitrile, 3-Amino-4-cyanopyrazole
Purity: 98%
CAS No.: 16617-46-2
Molecular Formula: C4H4N4
Formula Weight: 108.10
Odour: Odourless
Melting Point: 173-176°
Beilstein Reference: 2647
UN No.: UN3439
MDL No.: MFCD00005237
Formula:
C₄H₄N₄ Punto di fusione: 173…176 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00005237 CAS Number: 16617-46-2 EINECS: 240-665-6 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...