2-Phenylpropionitrile 96%
Fornitore: Thermo Fisher Scientific
|
Pericolo
|
Purity: 97%
CAS No.: 1823-91-2
Molecular Formula: C9H9N
Formula Weight: 131.18
Flash Point: 93°(199°F)
Refractive Index: 1.5106
Boiling Point: 230-232°
Beilstein Reference: 774576
UN No.: UN3276
MDL No.: MFCD00001871
Formula:
C₉H₉N Punto di ebollizione: 230…232 °C Densità: 0,985 Flash Pt: 93 °C (199 °F) Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00001871 CAS Number: 1823-91-2 EINECS: 217-354-9 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...