Stampa…

Reactive blue 4 (dye content) 35%, dark blue powder C.I. 61205

Fornitore: Thermo Scientific
Attenzione

A12911.14 A12911.22
A12911.14EA 43.4 EUR
A12911.14 A12911.22
Reactive blue 4 (dye content) 35%, dark blue powder C.I. 61205
Reactive blue 4

CAS No.: 13324-20-4 Molecular Formula: C23H14Cl2N6O8S2 Formula Weight: 637.44 Odour: Odourless MDL No.: MFCD00001218

Formula: C₂₃H₁₄Cl₂N₆O₈S₂
Peso molecolare: 637,44 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00001218
CAS Number: 13324-20-4
EINECS: 236-363-9

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR