Stampa…

4-Methylesculetin 97%

Fornitore: Thermo Scientific
Attenzione

Synonyms: 6,7-Dihydroxy-4-methylcoumarin, 6,7-Dihydroxy-4-methyl-2H-chromen-2-one

A11902.06 A11902.14 A11902.22
A11902.06EA 31.2 EUR
A11902.06 A11902.14 A11902.22
4-Methylesculetin 97%
4-Methylesculetin

Purity: 97% CAS No.: 529-84-0 Molecular Formula: C10H8O4 Formula Weight: 192.17 Melting Point: ca 275° dec. Beilstein Reference: 162550 MDL No.: MFCD00006859

Formula: C₁₀H₈O₄
Peso molecolare: 192,17 g/mol
Punto di fusione: ∼ 275 °C (dec.)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00006859
CAS Number: 529-84-0
EINECS: 208-470-0

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR