Stampa…

Benzil dioxime 98%

Fornitore: Thermo Fisher Scientific

A10687.14 A10687.22
A10687.14EA 61.4 EUR
A10687.14 A10687.22
Benzil dioxime 98%
Benzil dioxime

Purity: 98% CAS No.: 23873-81-6 Molecular Formula: C14H12N2O2 Formula Weight: 240.26 Melting Point: ca 238° dec. Beilstein Reference: 2053615 MDL No.: MFCD00002113

Formula: C₁₄H₁₂N₂O₂
Punto di fusione: ∼ 238 °C (dec.)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00002113
CAS Number: 23873-81-6
EINECS: 245-921-0
Merck Index: 14,1079

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR