Zeolite β, ammonium
Fornitore: Thermo Fisher Scientific
CAS No.: 1318-02-1
Molecular Formula: SiO2:Al2O3
Physical Form: Powder, S.A. 680 m2/g, 25:1 mole ratio
Appearance: White
MDL No.: MFCD00132601
Applications: Zeolite beta is used in isomerization of waxes, and Friedel Crafts reactions (alkylation and acylation)
Formula:
SiO₂:Al₂O₃ Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00132601 CAS Number: 1318-02-1 EINECS: 215-283-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...