Zeolite Y, sodium
Fornitore: Thermo Scientific
CAS No.: 1318-02-1
Molecular Formula: SiO2:Al2O3
Physical Form: Powder, S.A. 900 m2/g, 5.1:1 mole ratio
Appearance: White
MDL No.: MFCD00132601
Applications: Zeolite Y is used as a cracking catalyst in petroleum refinery catalytic cracking units to convert high-boiling fractions of petroleum crude to more valuable gasoline, diesel and other products
Formula:
SiO₂:Al₂O₃ Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00132601 CAS Number: 1318-02-1 EINECS: 215-283-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...