Povidone iodine
Fornitore: Thermo Scientific
Synonyms:
Polyvinylpyrrolidone-iodine complex
45782.A1
45782.18
45782.30
45782.A1EA
606
EUR
45782.A1
45782.18
45782.30
Povidone iodine
Povidone iodine
CAS No.: 25655-41-8
Molecular Formula: (C6H9NO)n.xI
Physical Form: Powder
Appearance: Yellow-brown
Melting Point: 300° dec.
MDL No.: MFCD00084483
Formula:
C₆H₉I₂NO Peso molecolare: 364,953 g/mol Punto di fusione: 300 °C (dec.) Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00084483 CAS Number: 25655-41-8 Merck Index: 14,7698 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...