Stampa…

Triethylenetetramine-N,N,N',N'',N''',N'''-hexaacetic acid ≥98%

Fornitore: Thermo Scientific
Attenzione

Synonyms: TTHA

44737.04 44737.09
44737.04EA 61.2 EUR
44737.04 44737.09
Triethylenetetramine-N,N,N',N'',N''',N'''-hexaacetic acid ≥98%
Triethylenetetramine-N,N,N',N'',N''',N'''-hexaacetic acid

Purity: 98% CAS No.: 869-52-3 Molecular Formula: C18H30N4O12 Formula Weight: 494.45 Physical Form: Powder MDL No.: MFCD00004290

Formula: C₁₈H₃₀N₄O₁₂
Peso molecolare: 494,45 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00004290
CAS Number: 869-52-3
EINECS: 212-790-6

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR