Humic acid ≥98%
Fornitore: Thermo Scientific
41747.06
41747.14
41747.06EA
49.5
EUR
41747.06
41747.14
Humic acid ≥98%
Humic acid
CAS No.: 1415-93-6
Physical Form: Crystalline powder
Appearance: Dark brown
Solubility: Slightly soluble in water, with much swelling
MDL No.: MFCD00147177
Applications: Metal chelator; pigments
Formula:
C₉H₉NO₆ Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00147177 CAS Number: 1415-93-6 EINECS: 215-809-6 Merck Index: 14,4752 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...