Stampa…

Lithium manganese(III,IV) oxide ≥99.5% (metals basis)

Fornitore: Thermo Scientific

40250.14 40250.22
40250.14EA 68.3 EUR
40250.14 40250.22
Lithium manganese(III,IV) oxide ≥99.5% (metals basis)
Lithium manganese(III,IV) oxide

Purity: 99.5% CAS No.: 12057-17-9 Molecular Formula: LiMn2O4 Formula Weight: 180.81 Physical Form: -325 Mesh Powder Odour: Odourless Melting Point: 400° MDL No.: MFCD01114233

Formula: LiMn₂O₄
Peso molecolare: 180,81 g/mol
Punto di fusione: >400 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD01114233
CAS Number: 12057-17-9

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR