Gold(I) sodium thiosulphate hydrate ≥99.9% (metals basis)
Fornitore: Thermo Scientific
Synonyms:
Sodium gold(I) thiosulphate hydrate
Purity: 99.9%
CAS No.: 15283-45-1
Molecular Formula: Na3Au(S2O3)2.xH2O
Formula Weight: 490.19(anhy)
Physical Form: Powder
Density: 3.09
Appearance: White
Melting Point: dec.
Solubility: Soluble in water. Insoluble in ethanol and most other organic solvents
MDL No.: MFCD00046176
Formula:
AuH₂Na₃O₆S₄·xH₂O Peso molecolare: 490,19 g/mol (anhy) Punto di fusione: Dec. Densità: 3,09 Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00046176 CAS Number: 15283-45-1 EINECS: 239-324-4 Merck Index: 14,4519 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...