Potassium isopropoxide 5% (w/v) in isopropanol (IPA) , 99% (metals basis)
Fornitore: Thermo Scientific
|
Pericolo
|
Purity: 99%
CAS No.: 6831-82-9
Molecular Formula: C3H7KO
Formula Weight: 98.19
Flash Point: 12°(54°F)
Physical Form: Liquid
Sensitivity: Air and Moisture Sensitive
Appearance: Colourless to pale yellow
Odour: Alcohol-like
UN No.: UN3274
MDL No.: MFCD00210641
Formula:
C₃H₇KO Flash Pt: 12 °C (54 °F) Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00210641 CAS Number: 6831-82-9 EINECS: 229-909-2 UN: 3274 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...