Tetrasodium meso-tetra(sulphonatophenyl)porphine dodecahydrate ≥95%
Fornitore: Thermo Scientific
|
Attenzione
|
Synonyms:
4,4',4'',4'''-(Porphine-5,10,15,20-tetrayl)tetrakis(benzene-sulphonic acid) tetrasodium salt dodecahydrate
Purity: 95%
CAS No.: 122451-08-5
Molecular Formula: C44H26N4Na4O12S4.12H2O
Formula Weight: 1239.11 (1022.87anhy)
Physical Form: Crystalline
Beilstein Reference: 5488862
MDL No.: MFCD00151011
Formula:
C₄₄H₂₆N₄Na₄O₁₂S₄·12H₂O Peso molecolare: 1239,11 g/mol (1022,87 anhy) Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00151011 CAS Number: 122451-08-5 EINECS: 254-262-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...