Stampa…
Ceramic tubes, one end closed
Fornitore: Thermo Scientific
30093.DR 30093.DW
30093.DREA 244 EUR
30093.DR 30093.DW
Ceramic tubes, one end closed
Tubazioni Tubazione di trasferimento
Pure Al₂O₃ Aluminium oxide AL24.
Slightly porous, good resistance to thermal shock, extremely good creep strength.