Stampa…

Alluminio ossido attivo acida Brockmann Grade I 58 angstroms

Fornitore: Thermo Fisher Scientific

11501.A1 11501.A7 11501.30
11501.A1EA 85.5 EUR
11501.A1 11501.A7 11501.30
Alluminio ossido attivo acida Brockmann Grade I 58 angstroms
Aluminium oxide activated acidic

CAS No.: 1344-28-1 Molecular Formula: Al2O3 Formula Weight: 101.96 Physical Form: -60 Mesh Powder, S.A. 150m2/g Odour: Odourless Melting Point: 2045° MDL No.: MFCD00003424


Suggested for the separation of both inorganic anions and acidic organic molecules such as acidic amino acids, aromatic acids and carboxylic acids

Formula: Al₂O₃
Peso molecolare: 101.96 g/mol
Densità: 3.94 g/cm³ (20 °C)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6
REACH: 01-2119529248-35
Merck Index: 14,356

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR