Iron(III) hydroxide, γ-phase
Fornitore: Thermo Scientific
17531.04
17531.09
17531.18
17531.04EA
146
EUR
17531.04
17531.09
17531.18
Iron(III) hydroxide, γ-phase
Iron(III) hydroxide, gamma-phase
CAS No.: 20344-49-4
Molecular Formula: FeO(OH)
Formula Weight: 88.85
Physical Form: -60 Mesh Powder
Odour: Odourless
Melting Point: 135° dec.
MDL No.: MFCD00064782
Formula:
FeO(OH) Peso molecolare: 88,85 g/mol Punto di fusione: 135 °C (dec.) Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00064782 CAS Number: 20344-49-4 EINECS: 243-746-4 Merck Index: 14,4025 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...