Ammonium aquopentachlororhodate(III) (≥30% Rh)
Fornitore: Thermo Fisher Scientific
|
Attenzione
|
CAS No.: 63771-33-5
Molecular Formula: (NH4)2[RhCl5(H2O)]
Formula Weight: 334.26
Physical Form: Powder
Melting Point: 210-230° dec.
MDL No.: MFCD00064673
Formula:
(NH₄)₂[RhCl₅(H₂O)] Peso molecolare: 334.26 g/mol Punto di fusione: 210…230 °C (dec.) Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00064673 CAS Number: 63771-33-5 EINECS: 264-455-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...