Potassium pentachlororuthenate(III) hydrate (≥25,4% Ru) ≥99.95% (metals basis)
Fornitore: Thermo Scientific
12173.03
12173.06
12173.03EA
91.9
EUR
12173.03
12173.06
Potassium pentachlororuthenate(III) hydrate (≥25,4% Ru) ≥99.95% (metals basis)
Potassium pentachlororuthenate(III) hydrate
Purity: 99.95%
Molecular Formula: K2RuCl5.xH2O
Formula Weight: 356.54(anhy)
Physical Form: Crystalline
MDL No.: MFCD00058747
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...