Chloropentaammineruthenium(III) chloride (≥33,5% Ru)
Fornitore: Thermo Fisher Scientific
|
Attenzione
|
CAS No.: 18532-87-1
Molecular Formula: Cl3H15N5Ru
Formula Weight: 292.58
Physical Form: Micro Crystals
Sensitivity: Hygroscopic
Appearance: Yellow/orange
MDL No.: MFCD00011529
Formula:
Cl₃H₁₅N₅Ru Peso molecolare: 292.58 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00011529 CAS Number: 18532-87-1 EINECS: 242-408-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...