Tungsten rhenium ≥99.95% (metals basis), in filo, Ø 0.25 mm (0.01 in)
Fornitore: Thermo Scientific
10073.G2
10073.G6
10073.G2EA
93.5
EUR
10073.G2
10073.G6
Tungsten rhenium ≥99.95% (metals basis), in filo, Ø 0.25 mm (0.01 in)
Tungsten rhenium
Purity: 99.95%
Molecular Formula: W:Re; 97:3 wt%
Physical Form: ≈1.9g/2m
MDL No.: MFCD02091751
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...