Stampa…
Mascherine per camera bianca, BioClean MTA™, PP
Fornitore: Nitritex
MTA210-0 MTA210-2
113-2251EA 584 EUR
113-2251 113-8794
Mascherine per camera bianca, BioClean MTA™, PP
Maschere Maschere per cleanroom
Providing high particle and bacterial filtration efficiency, the latex-free and low-linting BioClean MTA™ ensures protection, combined with comfort and exceptional breathability.
  • Three layers: Inner layer non-woven PP/PE (hygroscopic), filter layer meltblown PP, outer layer non-woven PP/PE (hydrophobic)
  • Ultra-low particulation
  • Latex-free
  • High strength seams, high BFE and PFE
  • Non-woven PP ties and plastic coated steel noseband
Adatte per camera bianca Classe 10, ISO 4 o M2.5.
Certificazioni: Manufactured in a facility holding ISO 9001:2015.More Product Information