Stampa…
Safety shoes, sandals, Velcro® fastening, Step One, Shuttle
Fornitore: U-Power
SO30025/36 SO30025/37 SO30025/38 SO30025/39 SO30025/40 SO30025/41 SO30025/42 SO30025/43 SO30025/44 SO30025/45 SO30025/46 SO30025/47 SO30025/48
UPOWSO30025/36EA 58.6 EUR
UPOWSO30025/36 UPOWSO30025/37 UPOWSO30025/38 UPOWSO30025/39 UPOWSO30025/40 UPOWSO30025/41 UPOWSO30025/42 UPOWSO30025/43 UPOWSO30025/44 UPOWSO30025/45 UPOWSO30025/46 UPOWSO30025/47 UPOWSO30025/48
Safety shoes, sandals, Velcro® fastening, Step One, Shuttle
Scarpe e stivali
Safety shoes made from leather and a slip-resistant dual-density PU sole.
  • Comfortable, anatomically shaped ergo dry insole features a dual PE/EVA support system with textile surface; anti-static and anti-fungal treated
  • Wingtex tunnel lining with honeycomb structure ensures breathability and excellent absorbance features
  • Stress-out tongue system allows easy instep and provides added comfort when walking
  • Natural fit width 11 for additional comfort
  • Air toe protective composite toecap with breathable membrane (200 J)
  • Puncture-resistant steel midsole
Colour: Black
Certificazioni: EN 20345:2011; S1P, SRC