Stampa…
Safety shoes, lace-up, The Roar, Joy
Fornitore: U-Power
SN20096/45 SN20096/47 SN20096/48 SN20096/44 SN20096/46 SN20096/35 SN20096/36 SN20096/37 SN20096/38 SN20096/39 SN20096/40 SN20096/41 SN20096/42 SN20096/43
UPOWSN20096/45EA 95.4 EUR
UPOWSN20096/45 UPOWSN20096/47 UPOWSN20096/48 UPOWSN20096/44 UPOWSN20096/46 UPOWSN20096/35 UPOWSN20096/36 UPOWSN20096/37 UPOWSN20096/38 UPOWSN20096/39 UPOWSN20096/40 UPOWSN20096/41 UPOWSN20096/42 UPOWSN20096/43
Safety shoes, lace-up, The Roar, Joy
Scarpe e stivali
Safety shoes made from velours leather and a slip-resistant dual-density PU sole.
  • Light non-slip Actiondry insole made from EVA/textile keeps feet dry; anti-fungal treated
  • Wingtex tunnel lining with honeycomb structure ensures breathability and excellent absorbance features
  • Natural fit width 11 for additional comfort
  • Airtoe protective aluminium toecap with breathable membrane (200 J)
  • Light, flexible and non-metallic puncture resistant midsole 'Save & Flex' covers 100% of the sole of the foot
Colour: Black
Colour: Black
Certificazioni: EN 20345:2011; S1P, SRC