Stampa…
Sleeve protectors, PE
Fornitore: Hubei Orient International
HOIT115390EA 161 EUR
HOIT115390 HOIT115391
Sleeve protectors, PE
Indumenti usa e getta
These disposable sleeve protectors made of PE provide waterproof protection for light-duty tasks.
  • Lightweight and soft
  • Waterproof and oil-proof
  • Breathable fabric
  • Latex-free
Ideal for use in non hazardous applications, such as cleaning, maintenance, food processing and handling, manufacturing, hotels and catering.