Stampa…
Tetremetilammonio idrossido 0.1 mol/L (0.1 N) in 2-propanol/methanol, Titripur® Reag. USP for titration in non-aqueous media, Supelco®
Fornitore: Merck
Pericolo
M-Clarity™ quality level = MQ100
1.08124.0250 1.08124.1000 1.08124.1003
1.08124.0250EA 107 EUR
1.08124.0250 1.08124.1000 1.08124.1003
Tetremetilammonio idrossido 0.1 mol/L (0.1 N) in 2-propanol/methanol, Titripur® Reag. USP for titration in non-aqueous media, Supelco®
Tetremetilammonio idrossido
Merck have introduced into the range of Titripur® Titrants some packs with additional technology - Supelco® SmartTitrants that use an RFID tag to provide a seamless data transfer between the smart titrator and the RFID tag on the label of the bottle of titrant. It will ensure that all information needed for method execution, result calculation and documentation is transferred to the titrator equipped with this technology. MettlerToledo Compact and Excellence titrators include this technology. Please see ordering table below for packs with RFID tag.
Formula: C₄H₁₃NO
Peso molecolare: 91.15 g/mol
Densità: 0.79 g/cm3 (20 °C)
Flash Pt: 15 °C
MDL Number: MFCD00008280
CAS Number: 75-59-2
UN: 1992
ADR: 3 (6.1) II
Merck Index: 14,09224
Visualizza i codici
Form liquid
Amount-of-substance concentration 0.0995 - 0.1005
Measurement uncertainty +/- 0.0003
Traceability NIST SRM
The concentration is determined by volumetric titration and refers to 20°C.