Stampa…
VWR®, Test Sieves, 450×70 mm, with Square Holes
Fornitore: VWR Collection
510-1661EA 562 EUR
510-1661 510-1668 510-1667 510-1666 510-1665 510-1664 510-1662 510-1671 510-1660 510-1659 510-1658 510-1657 510-1656 510-1655 510-1663 510-1677 510-1684 510-1683 510-1682 510-1681 510-1680 510-1669 510-1678 510-1670 510-1676 510-1675 510-1674 510-1673 510-1672 510-1679
VWR®, Test Sieves, 450×70 mm, with Square Holes
Setacci Setacci per analisi
Frame: Stainless steel; perforated plates: Galvanised steel, ISO 3310-2, ASTM E323, BS 410, AFNOR.
Optional: Inspection certificate of calibration 3.1 according to DIN EN 10204 with measuring results of a specific test.
Sieves and accessories of other dimensions are available on request.
Please go to website for a full overview.
All sieves are supplied with serial number and a certificate of compliance with the order 2.1 according to DIN EN 10204. Further certificates and calibration services are available. Contact your local Avantor sales office for further details.
Informazioni per l'ordine: Supplied as standard with a certificate of compliance 2.1 according to DIN EN 10204.