Stampa…
VWR®, Test Sieves, 250×55 mm, with Square Holes
Fornitore: VWR Collection
510-1046EA 263 EUR
510-1046 510-1047 510-1048 510-1068 510-1069 510-1070 510-1071 510-1072 510-1073 510-1074 510-1050 510-1051 510-1052 510-1053 510-1054 510-1075 510-1076 510-1077 510-1078 510-1079 510-1080 510-1081 510-1082 510-1083 510-1084 510-1085 510-1049 510-1086 510-1055 510-1056 510-1057 510-1058 510-1059 510-1060 510-1061 510-1062 510-1063 510-1064 510-1065 510-1066 510-1067
VWR®, Test Sieves, 250×55 mm, with Square Holes
Setacci Setacci per analisi
Frame: Stainless steel; perforated plates: Galvanised steel, ISO 3310-2, ASTM E323, BS 410, AFNOR.
Optional: Inspection certificate of calibration 3.1 according to DIN EN 10204 with measuring results of a specific test.
Sieves and accessories of other dimensions are available on request.
Please go to website for a full overview.
All sieves are supplied with serial number and a certificate of compliance with the order 2.1 according to DIN EN 10204. Further certificates and calibration services are available. Contact your local Avantor sales office for further details.
Informazioni per l'ordine: Supplied as standard with a certificate of compliance 2.1 according to DIN EN 10204.