Stampa…
VWR®, Setacci per analisi, 203×50 mm
Fornitore: VWR Collection
510-0911EA 793 EUR
510-0911 510-0912 510-0913 510-0914 510-0915 510-0916 510-0917 510-0918 510-0919 510-0920 510-0921 510-0922 510-0923 510-0924 510-0925 510-0926 510-0927 510-0928 510-0929 510-0930 510-0931 510-0932 510-0933 510-0934 510-0935 510-0936 510-0937 510-0938 510-0939 510-0940 510-2500 510-2501 510-2502 510-2503 510-2504 510-2505 510-2506 510-2507 510-2508 510-2509 510-2510 510-2511 510-2512 510-2513 510-2514 510-2515 510-2516 510-2517 510-2518 510-2519 510-2520 510-2521 510-2522 510-2523 510-2524 510-2525 510-2526 510-2566 510-2567 510-2568 510-2527 510-2528 510-2529 510-2530 510-2531 510-2532 510-2533 510-2534 510-2535 510-2536 510-2537 510-2538 510-2539 510-2540 510-2541 510-2542 510-2543 510-2544 510-2545 510-2546 510-2547 510-2548 510-2549 510-2550 510-2551 510-2552 510-2553 510-2554 510-2555 510-2556 510-2557 510-2558 510-2559 510-2560 510-2561 510-2562 510-2563 510-2564 510-2565
VWR®, Setacci per analisi, 203×50 mm
Setacci Setacci per analisi
Frame: Stainless steel; woven wire cloth: Stainless steel, ISO 3310, ASTM E11, BS 410, AFNOR.
Optional: Inspection certificate of calibration 3.1 according to DIN EN 10204 with measuring results of a specific test.
Sieves and accessories of other dimensions are available on request.
Please go to website for a full overview.
All sieves are supplied with serial number and a certificate of compliance with the order 2.1 according to DIN EN 10204. Further certificates and calibration services are available. Contact your local Avantor sales office for further details.
Informazioni per l'ordine: Supplied as standard with a certificate of compliance 2.1 according to DIN EN 10204.