Stampa…
Wafer Storage Boxes for 150 mm Wafer Handling, E 90, E 124 and E 160 Series
Fornitore: ENTEGRIS
E16060103S69F2 E1246010161C02 E124601010603 E1606010369F02 E90-101-61C02 E1246010169F02 E124601010604 E90-101-0603
ENTEE16060103S69F2EA 0 EUR
ENTEE16060103S69F2 ENTEE1246010161C02 ENTEE124601010603 ENTEE1606010369F02 ENTEE90-101-61C02 ENTEE1246010169F02 ENTEE124601010604 ENTEE90-101-0603
Wafer Storage Boxes for 150 mm Wafer Handling, E 90, E 124 and E 160 Series
Manipolazione dei wafer Scatole per lo stoccaggio dei wafer
E 90 series
  • Maximum stacking density for space limited stockers and WIP racks.
  • Smooth bottom
  • Pin and socket hinges
  • Stackable
  • Single carrier capacity
  • Compatible carriers: 182 series and 194/196 series 25-capacity without handles
E 124 series
The industry standard 150 mm storage box
Automation compatible
Smooth surfaces for easy cleaning and low particle generation
Drop front allows easy access by robotic end effectors
Robotic compatible slide latch
Rotating pin and socket hinges prevent box from tipping when open
Stackable
Single carrier capacity
Compatible carriers: All 25-capacity without handles
E 160 series
Optimum performance and reduced particle generation.
Five degree wafer tilt from wafers vertical allows wafers to rest on their backsides, reducing movement and particle generation
Smooth surfaces for easy cleaning and low particle generation
Pin and socket hinges with retainers for improved durability
Stackable
Single carrier capacity
Compatible carriers: All 25-capacity without handles