Stampa…
Bottiglie a collo stretto, quadrate, PC, senza tappi a vite, Nalgene®
Fornitore: Thermo Fisher Scientific
362015-0125 362015-0250 362015-0500 362015-1000
NALG362015-0125EA 0 EUR
NALG362015-0125 NALG362015-0250 NALG362015-0500 215-7000
Bottiglie a collo stretto, quadrate, PC, senza tappi a vite, Nalgene®
Bottiglie Bottiglie in plastica
PC, transparent, without screw cap.
  • Easy-to-handle square design
  • Tough, durable even at low temperatures
  • With molded-in graduations and drip-proof neck
Ideal for storage of aqueous solutions and short-term storage of media.
These bottles are autoclavable.