Stampa…
Anti-Heat Shock Protein 70 Mouse Monoclonal Antibody [clone: W27]
Fornitore: US Biological
H1830-95V2-25 H1830-95V2-100
USBIH1830-95V2-25EA 343 EUR
USBIH1830-95V2-25 USBIH1830-95V2-100
Anti-Heat Shock Protein 70 Mouse Monoclonal Antibody [clone: W27]
Anticorpi
Recommended dilution: Western Blot: 0.25-1 ?g/ml
Type: Primary
Antigen: Heat Shock Protein 70
Clonality: Monoclonal
Clone: W27
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human, Mouse,