Stampa…

Anti-Cyclin B1 Mouse Monoclonal Antibody [clone: V152]

Fornitore: US Biological
C8453-04F-25 C8453-04F-100
USBIC8453-04F-25EA 405 EUR
USBIC8453-04F-25 USBIC8453-04F-100
Anti-Cyclin B1 Mouse Monoclonal Antibody [clone: V152]
Anticorpi
Recommended dilution: Western Blot: 2.5-5 ?g/5ml

Type: Primary
Antigen: Cyclin B1
Clonality: Monoclonal
Clone: V152
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse,
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR