Stampa…

Anti-Myc-Tag Mouse Monoclonal Antibody (Agarose) [clone: 10K114]

Fornitore: US Biological
C0035-07N1-5 C0035-07N1-500
USBIC0035-07N1-5EA 1140 EUR
USBIC0035-07N1-5 USBIC0035-07N1-500
Anti-Myc-Tag Mouse Monoclonal Antibody (Agarose) [clone: 10K114]
Anticorpi
Type: Primary
Antigen: Myc-Tag
Clonality: Monoclonal
Clone: 10K114
Conjugation: Agarose
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human,
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR