Stampa…
Anti-Hemagglutinin Tag Mouse Monoclonal Antibody (Agarose) [clone: 10K238]
Fornitore: US Biological
H1847-54Q-500 H1847-54Q-5
USBIH1847-54Q-500EA 387 EUR
USBIH1847-54Q-500 USBIH1847-54Q-5
Anti-Hemagglutinin Tag Mouse Monoclonal Antibody (Agarose) [clone: 10K238]
Anticorpi
Type: Tag
Antigen: Hemagglutinin Tag
Clonality: Monoclonal
Clone: 10K238
Conjugation: Agarose
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human,